missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation ACQUITY BEH Phenyl Column

ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds.

Brand:  Waters Corporation 176000905

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10645916

  • £3000.00 / Pack of 3
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

  • This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemically stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Specifications

Specifications

1 mm
18000 psi (1240 Bar)
1.7 μm
3/Pk.
Yes
50 mm
Spherical
130 Å
L11
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers