missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACK1 Antibody (ACK 1F4/9), Novus Biologicals™
Mouse Monoclonal Antibody
£149.00 - £301.00
Specifications
| Antigen | ACK1 |
|---|---|
| Clone | ACK 1F4/9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688078
|
Novus Biologicals
NBP2-50100-0.025ml |
0.025 mL |
£149.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635467
|
Novus Biologicals
NBP2-50100 |
0.1 mL |
£301.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACK1 Monoclonal antibody specifically detects ACK1 in Drosophila samples. It is validated for Western Blot, ELISASpecifications
| ACK1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Protein Kinase, Signal Transduction | |
| PBS | |
| 10188 | |
| IgM | |
| Thiophilic Chromatography |
| ACK 1F4/9 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Drosophila | |
| ACK-1, ACK1ACKp21cdc42Hs, activated CDC42 kinase 1, activated Cdc42-associated kinase 1, activated p21cdc42Hs kinase, EC 2.7.10, EC 2.7.10.2, FLJ44758, FLJ45547, Tyrosine kinase non-receptor protein 2, tyrosine kinase, non-receptor, 2 | |
| ACK peptides (drosophila) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title