missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACK1 Antibody (ACK 1F4/9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50100
This item is not returnable.
View return policy
Description
ACK1 Monoclonal antibody specifically detects ACK1 in Drosophila samples. It is validated for Western Blot, ELISA
Specifications
| ACK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| ACK-1, ACK1ACKp21cdc42Hs, activated CDC42 kinase 1, activated Cdc42-associated kinase 1, activated p21cdc42Hs kinase, EC 2.7.10, EC 2.7.10.2, FLJ44758, FLJ45547, Tyrosine kinase non-receptor protein 2, tyrosine kinase, non-receptor, 2 | |
| ACK peptides (drosophila) | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 10188 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, ELISA | |
| ACK 1F4/9 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| Drosophila | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction