missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £309.00
Specifications
| Antigen | Acinus |
|---|---|
| Dilution | Western Blot 1:500 -1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263551
|
Novus Biologicals
NB500-216SS |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18266920
|
Novus Biologicals
NB500-216 |
0.1 mg |
£309.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot.Specifications
| Acinus | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| 22985 | |
| C-terminus of Acinus (proprietary peptide). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500 -1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
| ACIN1 | |
| IgG | |
| IgG purified | |
| Reacts with the N terminus of Acinus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title