missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-216
This item is not returnable.
View return policy
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot.
Specifications
| Acinus | |
| Polyclonal | |
| Western Blot 1:500 -1:1000 | |
| Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Reacts with the N terminus of Acinus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ACIN1 | |
| C-terminus of Acinus (proprietary peptide). | |
| 0.1 mg | |
| Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
| 22985 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction