missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00
Specifications
| Antigen | Acinus |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Acinus | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
| ACIN1 | |
| IgG | |
| Affinity Purified | |
| 220 kDa |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAD56724 | |
| 22985 | |
| Antibody was raised against a 35 amino acid peptide near the center of human Acinus. The immunogen is located within amino acids 760 - 810 of Acinus. Amino Acid Squence: DTSENRPENDVPEPP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title