missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76855-0.025mg
This item is not returnable.
View return policy
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Acinus | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
| Rabbit | |
| 220 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
| 22985 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5-20 μg/mL | |
| AAD56724 | |
| ACIN1 | |
| Antibody was raised against a 35 amino acid peptide near the center of human Acinus. The immunogen is located within amino acids 760 - 810 of Acinus. Amino Acid Squence: DTSENRPENDVPEPP | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction