missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-Lysine Antibody (1C6), Novus Biologicals™
Mouse Monoclonal Antibody
£169.00
Specifications
| Antigen | Acetyl-Lysine |
|---|---|
| Clone | 1C6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 3 μg, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 10 μg |
| Applications | Western Blot |
Description
Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot, ChIP assay.Specifications
| Acetyl-Lysine | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| Synthetic peptide sequence surrounding the acetylated lysine 9 of histone H3. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1C6 | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 3 μg, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 10 μg | |
| Monoclonal | |
| Purified | |
| RUO | |
| acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine | |
| IgG1 | |
| Protein G purified | |
| Acetylated Lysine (1C6) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title