missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-Lysine Antibody (1C6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-74339-0.025mg
This item is not returnable.
View return policy
Description
Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot, ChIP assay.
Specifications
| Acetyl-Lysine | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 3 μg, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 10 μg | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Acetylated Lysine (1C6) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1C6 | |
| Unconjugated | |
| acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine | |
| Synthetic peptide sequence surrounding the acetylated lysine 9 of histone H3. | |
| 0.025 mg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction