missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA, p Ser79 Antibody (SR1413), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21598-100ul
This item is not returnable.
View return policy
Description
Acetyl-CoA Carboxylase alpha/ACACA Monoclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Acetyl-CoA Carboxylase alpha/ACACA | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1413 | |
| Western Blot 1:500-1:2000 | |
| ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
| A synthesized peptide derived from human phospho-Acetyl-CoA Carboxylase alpha/ACACA [Ser79] (Uniprot #: Q13085) | |
| 100 μg | |
| Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
| 31 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction