missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-CoA Carboxylase alpha/ACACA, p Ser79 Antibody (SR1413), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | Acetyl-CoA Carboxylase alpha/ACACA |
|---|---|
| Clone | SR1413 |
| Dilution | Western Blot 1:500-1:2000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643675
|
Novus Biologicals
NBP3-21598-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617964
|
Novus Biologicals
NBP3-21598-50ul |
50 μL |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetyl-CoA Carboxylase alpha/ACACA Monoclonal antibody specifically detects Acetyl-CoA Carboxylase alpha/ACACA in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Acetyl-CoA Carboxylase alpha/ACACA | |
| Western Blot 1:500-1:2000 | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Lipid and Metabolism, Neuroscience, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 31 | |
| Affinity purified |
| SR1413 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ACACacetyl-CoA carboxylase 1, ACC1ACC, ACCA, ACC-alpha, acetyl-CoA carboxylase alpha, acetyl-Coenzyme A carboxylase alpha, EC 6.4.1.2 | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title