missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™

£212.00 - £453.00

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10398233
View Documents Promotion Details
Fisher Chemical
A/0632/PB15
1 L
£212.00
1 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10629112
View Documents Promotion Details
Fisher Chemical
A/0632/PB17
2.5 L
£453.00
2.50 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acetonitrile, Advanced HPLC Grade for HPLC Gradient Analysis

Specifications

Chemical Identifiers

75-05-8
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
CC#N
C2H3N
1878
acetonitrile

Specifications

-46°C
Colorless
Plastic-coated Glass Bottle
C2H3N
97mbar at 20°C
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
41.05g/mol
75-05-8
81°C
Liquid
1878
0.36cp at 20°C
CC#N
41.05
Videos
SDS
Documents

Documents

Product Certifications