missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid-2-13C, TRC

Product Code. 30178978
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178978 250 mg 250mg
30179223 1 g 1g
30178966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30178978 Supplier Toronto Research Chemicals Supplier No. TRCA167641250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1563-80-0
Molecular Formula C13CH4O2
Molecular Weight (g/mol) 61.04
Synonym [2-13C]Acetic Acid;
IUPAC Name acetic acid
SMILES [13CH3]C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1+1
Formula Weight 61.0245
Chemical Name or Material Acetic Acid-2-13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.