CAS RN 1563-80-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acetic Acid-2-13C, TRC
CAS: 1563-80-0 Molecular Formula: C13CH4O2 Molecular Weight (g/mol): 61.04 Synonym: [2-13C]Acetic Acid; IUPAC Name: acetic acid SMILES: [13CH3]C(=O)O