Learn More
Sperian™ Polycarbonate Visor
Product Code. 11841781
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Pack of 10
Product Code. Lens Material unitSize
11841781 Polycarbonate Pack of 10
1 options
Product Code. 11841781 Supplier Sperian™ Supplier No. 1002328
Only null left
Add to basket
Add to basket
Specifications
Certifications/Compliance 89/686/EEC, EN166, ISO 9001 / 2000
Lens Tint Shade 1.7, Green
Product Type Electrical Visor
Model SV9PG
Material Polycarbonate
Lens Material Polycarbonate