missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ACE-2 Inhibitor Screening Assay Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
96 Tests
Unit Size:
1 set
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Gene ID (Entrez) | 59272 |
| Gene Symbol | ACE2 |
| Quantity | 96 Tests |
| Synonym | ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 |
| Research Category | Immunology, Virology Bacteria and Parasites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction