missing translation for 'onlineSavingsMsg'
Learn More

ACE-2 Antibody (CL4013), Novus Biologicals™

Mouse Monoclonal Antibody

£292.00 - £435.00

Specifications

Antigen ACE-2
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:10000 - 1:20000, Immunohistochemistry-Paraffin 1:10000 - 1:20000
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18280394
View Documents
Novus Biologicals
NBP2-59036
100 μL
£435.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18608156
View Documents
Novus Biologicals
NBP2-59036-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACE-2 Monoclonal specifically detects ACE-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ACE-2
Monoclonal
Purified
RUO
Human
59272
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MSSSSWLLLSLVAVTAAQSTIEEQAKTFLDKFNHEAEDLFYQSSLASWNYNTNITEENVQNMNNAGDKWSAFLKEQSTLAQMYPLQEIQNLTVKLQLQALQQNGSSVLSED
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:10000 - 1:20000, Immunohistochemistry-Paraffin 1:10000 - 1:20000
Unconjugated
Mouse
Immunology, Virology Bacteria and Parasites
ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
ACE2
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.