Learn More
Fisherbrand™ accumet™ pH Indicating Epoxy Body Glass Membrane Models - Mercury - Free
Product Code. 11570094 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
U.S. Standard
Unit Size:
Each
Product Code. Connector Types unitSize
11570094 U.S. Standard Each
1 options
Product Code. 11570094
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Universal glass membrane and epoxy body provides measurements from zero to 14pH
  • Epoxy body
  • Universal glass membrane
  • Zero to 14pH range
TRUSTED_SUSTAINABILITY
Specifications
Connector Types U.S. Standard
Material Epoxy
pH Range 0 to 14
Type pH Membrane Model