missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free

Product Code. 11560174 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Connector Types:
BNC
U.S. Standard/Pin
Junction:
Double Ceramic
Single Ceramic
Filling Solution Type:
SP135-500
SP138-500
Length (Metric) Cable:
76.2 cm
152.4 cm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Junction Filling Solution Type Length (Metric) Cable Unit Size
11560174 BNC Double Ceramic SP138-500 76.2 cm Each
10232781 BNC Single Ceramic SP135-500 76.2 cm Each
11520064 U.S. Standard/Pin Single Ceramic SP135-500 76.2 cm Each
11530064 U.S. Standard/Pin Single Ceramic SP135-500 152.4 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11560174
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Removable bulb protector

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

  • 0 to 14 pH range
  • Porous ceramic junction
  • Twist-open/twist-close cap for easy filling
  • Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
  • Immersion depth, 4 in. (10.1cm)
  • 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead

Choice of Ag/AgCl Single- or Double-Junction internal reference elements

  • Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
  • Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
  • For aqueous and nonaqueous samples
  • Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers
TRUSTED_SUSTAINABILITY

Specifications

Closure Type Twist Open/Twist Close
Color Purple
Connector Types BNC
Junction Double Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating -5°C to +100°C
Type Combination pH Electrode
Filling Solution Type SP138-500
For Use With (Application) For aqueous and nonaqueous samples
Internal Reference Ag/AgCl
Temperature (English) Operating 23°F to 212°F
Temperature (Metric) Range -5°C to +100°C
Length (English) 4 in.
Length (English) Cable 30 in.
Length (Metric) 101.6 mm
Length (Metric) Cable 76.2 cm
Diameter (English) 0.4 in.
Diameter (Metric) 10 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.