missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AB150 Benchtop pH Meters

Product Code. 12840633 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Meter with BioBasic Kit
Meter with pH Kit
pH Meter
Unit Size:
Each
Product Code. Type unitSize
12840633 pH Meter Each
12880633 Meter with BioBasic Kit Each
12870633 Meter with pH Kit Each
3 options
This item is not returnable. View return policy

Product Code. 12840633

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

accumet AB150 pH benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply

Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.

  • Graphic backlit display with date and time, stability indicator and alarms
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • One point relative mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings

NIST-Traceable Calibration Models (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory; includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz

Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial

Certifications

Select models available with Certificate of NIST Traceability

Warranty

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Includes Meter, Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply
Accuracy (Temperature) ±0.3°C
Accuracy (mV) ±0.2
Accuracy (pH) ±0.002
Data Management Details Manual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format
Outputs RS-232 (phono plug), mini-B USB, stirrer
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Calibration Points (pH) Up to 6 pre-set or 5 custom
Range (mV) ±2000
Resolution (mV) 0.1 mV
Calibration Points 1 to 6
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Data Points 500
Inputs BNC, ATC
Certifications/Compliance Without NIST-traceable pH Certificate
Warranty 3 Years
Model AB150 pH Meter
Type pH Meter
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.