missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACCN2 Antibody (S271-44), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00
Specifications
| Antigen | ACCN2 |
|---|---|
| Clone | S271-44 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
ACCN2 Monoclonal specifically detects ACCN2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| ACCN2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat | |
| Acid-sensing ion channel 1, acid-sensing ion channel 1a protein, amiloride-sensitive cation channel 2, neuronal, ASIC, ASIC1ASIC1A, BNAC2, BNaC2ACCN2 variant 3, Brain sodium channel 2, Cation channel, amiloride-sensitive, neuronal, 2, hBNaC2 | |
| ASIC1 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 60kDa. |
| S271-44 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q6NXK8 | |
| 41 | |
| Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title