missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACCN2 Antibody (S271-44), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22409-0.025mg
This item is not returnable.
View return policy
Description
ACCN2 Monoclonal specifically detects ACCN2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ACCN2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Acid-sensing ion channel 1, acid-sensing ion channel 1a protein, amiloride-sensitive cation channel 2, neuronal, ASIC, ASIC1ASIC1A, BNAC2, BNaC2ACCN2 variant 3, Brain sodium channel 2, Cation channel, amiloride-sensitive, neuronal, 2, hBNaC2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 41 | |
| Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S271-44 | |
| Unconjugated | |
| Q6NXK8 | |
| ASIC1 | |
| Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1 | |
| 0.025 mg | |
| Primary | |
| Detects approx 60kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction