missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers

Product Code. p-8442004 Shop All Thermo Scientific Products
Click to view available options
Type:
Liquid Filter
Storage Bottle Cap
For Use With (Equipment):
Wellwash Microplate Strip Washer
Unit Size:
1 set
Pack of 2
This item is not returnable. View return policy

Product Code. 15640845

Brand: Thermo Scientific™ N12407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.

These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.

Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 Pk.
Type Liquid Filter
For Use With (Equipment) Wellwash Microplate Strip Washer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.