Learn More
Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.
Brand: Thermo Scientific™ N06210
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.
Varioskan LUX is a modular instrument, you can upgrade it with missing features later.- Installation of the upgrade kits must be carried out by trained and authorized service personnel.
- For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
- TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
- Please contact your local representative to discuss about the instrument upgrade option
- Adapters adjust the plates to the optimum height for measuring and dispensing.
- Choose the adapter based on the plate format in use and whether the lid is needed or not.
- The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
- A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
Specifications
1536-well adapter for 10 mm plate without lid | |
Varioskan LUX Microplate Reader | |
1536-well Plate | |
Varioskan™ |