missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Accessories for Microplate Reader

Product Code. 11573505 Shop All Agilent BioTek Products
Click to view available options
Product Type:
Fluorescence Filter
Serial Cable; Modem (9-pin female to 9-pin male)
Serial Cable; Null Modem (9-pin female to 25-pin female)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11573505

Brand: Agilent BioTek 75053

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Multi-mode, absorbance and monochromator based microplate readers.

Serial Cable (BT75034)

  • Modem (9-pin female to 9-pin male)
  • For use with ELx800, Synergy 2, Synergy 2 Alpha, Synergy 2 SL, Synergy H1m, Synergy HT, Synergy Mx, Synergy MxF
Serial Cable (BT75053)
  • Null Modem (9-pin female to 25-pin female)
  • For use with ELx808, FLx800, PowerWave, Precision, Precision XS, Synergy HT
Fluorescence Filter (BT7082225)
  • 590/20 nm
  • For use with Cytation3, FLx800, Synergy 2, Synergy H1, Synergy H4, Synergy HT
Fuse (BT46024)
  • 250V 1.5A
  • For use with ELx808
Fuse (BT46023)
  • 0.75A
  • For use with PowerWave X

TRUSTED_SUSTAINABILITY

Specifications

Product Type Serial Cable; Null Modem (9-pin female to 25-pin female)
For Use With (Equipment) Agilent BioTek Microplate Readers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.