missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Accessories for Microplate Reader
Shop All Agilent BioTek Products
Click to view available options
Product Type:
Fluorescence Filter
Serial Cable; Modem (9-pin female to 9-pin male)
Serial Cable; Null Modem (9-pin female to 25-pin female)
Unit Size:
Each
Description
Serial Cable (BT75034)
- Modem (9-pin female to 9-pin male)
- For use with ELx800, Synergy 2, Synergy 2 Alpha, Synergy 2 SL, Synergy H1m, Synergy HT, Synergy Mx, Synergy MxF
- Null Modem (9-pin female to 25-pin female)
- For use with ELx808, FLx800, PowerWave, Precision, Precision XS, Synergy HT
- 590/20 nm
- For use with Cytation3, FLx800, Synergy 2, Synergy H1, Synergy H4, Synergy HT
- 250V 1.5A
- For use with ELx808
- 0.75A
- For use with PowerWave X
Specifications
Specifications
| Product Type | Serial Cable; Modem (9-pin female to 9-pin male) |
| For Use With (Equipment) | Agilent BioTek Microplate Readers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction