Learn More
Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers
Optimize the Accessories for Thermo Scientific™ MaxQ™ 7000 Water Bath Shakers to increase the versatility of the MaxQ 7000.
Brand: Thermo Scientific™ SHK7000-30
Code : 50
Additional Details : Weight : 0.01000kg
Description
Put Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers to use to increase the utility of MaxQ 7000 Water Bath Shakers. With exceptional innovation, reliability and accuracy, Thermo Scientific MaxQ 7000 Shakers, Platforms and accessories are the optimal choice for your busy lab.
Specifications
Gable Cover | |
Stainless Steel | |
Stainless-steel gable cover for temperatures up to 90°C |