missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers
Optimize the Accessories for Thermo Scientific™ MaxQ™ 7000 Water Bath Shakers to increase the versatility of the MaxQ 7000.
£236.63 - £894.91
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10306732
|
Thermo Scientific™
SHK7000-30 |
Stainless-steel gable cover for temperatures up to 90°C |
£598.04
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
10071612
|
Thermo Scientific™
SHK7000-32 |
Plexiglas™ gable cover for temperatures up to 65°C |
£652.25
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
10245902
|
Thermo Scientific™
SHK7000-37 |
Cooling coil for near-ambient temperature and below (requires a circulating chiller) |
£247.82
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10001632
|
Thermo Scientific™
SHK7000-40 |
Auxiliary platform suspension arms for rapid platform changes |
£236.63
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10530293
|
Thermo Scientific™
SHK7000-45 |
Movable dolly for underLab bench operation |
£894.91
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
Put Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers to use to increase the utility of MaxQ 7000 Water Bath Shakers. With exceptional innovation, reliability and accuracy, Thermo Scientific MaxQ 7000 Shakers, Platforms and accessories are the optimal choice for your busy lab.