Learn More
Agilent BioTek Accessories for Lionheart FX Automated Live Cell Imager
Shop All Agilent BioTek Products
Click to view available options
Product Type:
Aligned injector tip
Environment Control Cover
Humidity Chamber with Rapid Gas Recharge
Injector Tubing Bracket
Microplate/Slide Stage Insert
Offset injector tip
Spare Oil
T25 Flask Adapter
T75 U-shaped flask insert
T75 flask stage insert
Unit Size:
Each
Description
Injector accessories with offset tips, for use with Lionheart FX. Comes with tip mount, tubing, priming trough and tubing bracket.
Specifications
Specifications
| Product Type | T75 flask stage insert |
| For Use With (Equipment) | Lionheart FX Automated Live Cell Imager |