Learn More
Agilent BioTek Accessories for Lionheart FX Automated Live Cell Imager
GREENER_CHOICE
Product Code. 16502152 Shop All Agilent BioTek Products
Click to view available options
Product Type:
Aligned injector tip
Environment Control Cover
Humidity Chamber with Rapid Gas Recharge
Injector Tubing Bracket
Microplate/Slide Stage Insert
Offset injector tip
Spare Oil
T25 Flask Adapter
T75 U-shaped flask insert
T75 flask stage insert
Unit Size:
Each
Product Code. 16502152
Brand: Agilent BioTek 1450009
Miscellaneous accessories for use with Lionheart FX Automated Live Cell Imager.
Injector accessories with offset tips, for use with Lionheart FX. Comes with tip mount, tubing, priming trough and tubing bracket.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Environment Control Cover
For Use With (Equipment) Lionheart FX Automated Live Cell Imager