Learn More
Sartorius Accessories for Combisart™ Systems: Filtrate Flasks
Filtrate flasks for use with Sartorius Combisart Systems
Brand: Sartorius 16672-----1
Code : NEW
Additional Details : Weight : 0.77000kg
Description
Filtrate flasks for use with Sartorius Combisart Systems
- Suction flask for individual filter holders
- Vacuum-resistant borosilicate glass, 3.3 (Duran 50 glass)
- Plastic hose nipple acc. to DIN 12476
- Stopper/tube
- Sterilizable
Specifications
Suction Flask | |
Translucent | |
For collecting the filtrate | |
Plastic hose nipple |
5000mL | |
Borosilicate Glass | |
Individual vacuum filter holder |