missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD6 Antibody (07), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | ACBD6 |
|---|---|
| Clone | 07 |
| Applications | ELISA, Sandwich ELISA Capture |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684608
|
Novus Biologicals
NBP3-06598-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603858
|
Novus Biologicals
NBP3-06598-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACBD6 Monoclonal antibody specifically detects ACBD6 in Human samples. It is validated for ELISA, Sandwich ELISA CaptureSpecifications
| ACBD6 | |
| ELISA, Sandwich ELISA Capture | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 84320 | |
| IgG | |
| Protein A purified |
| 07 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACBD6 (Accession#: NP_115736.1; Met1-Ala282). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title