missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD6 Antibody (07), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06598-50ul
This item is not returnable.
View return policy
Description
ACBD6 Monoclonal antibody specifically detects ACBD6 in Human samples. It is validated for ELISA, Sandwich ELISA Capture
Specifications
| ACBD6 | |
| Monoclonal | |
| Unconjugated | |
| acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACBD6 (Accession#: NP_115736.1; Met1-Ala282). | |
| 50 μg | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 84320 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Sandwich ELISA Capture | |
| 07 | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction