missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£171.00 - £372.00
Specifications
| Antigen | ACAT |
|---|---|
| Dilution | Western Blot 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 2-10 ug/ml lysate |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280090
|
Novus Biologicals
NB400-141SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756843
|
Novus Biologicals
NB400-141 |
0.1 mL |
£372.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACAT Polyclonal specifically detects ACAT in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| ACAT | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Hamster | |
| P35610 | |
| 6646 | |
| A fusion protein construct that contains the N-terminal region of human ACAT1. [UniProt# P35610] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 46 kDa |
| Western Blot 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 2-10 ug/ml lysate | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antisera with 0.02% Sodium Azide | |
| ACACT, ACACT1, ACAT-1, ACATACAT1, acyl-Coenzyme A: cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase 1, Cholesterol acyltransferase 1, EC 2.3.1.26, RP11-215I23.2, STATSOAT, sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1, sterol O-acyltransferase 1 | |
| SOAT1 | |
| IgG | |
| Unpurified | |
| This product is specific for protein corresponding to ACAT1 - Sterol O-acyltransferase 1 (SOAT1) Uniprot# P35610, Gene ID# 6646. [Important Note:- ACAT1 is a symbol for both acyl-Coenzyme A: cholesterol acyltransferase 1 (SOAT1/ACAT1) and for acetyl-CoA acetyltransferase 1 (ACAT1/ THIL, Gene ID 38, Uniprot P24752) which often cause confusion]. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title