missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB400-141SS
This item is not returnable.
View return policy
Description
ACAT Polyclonal specifically detects ACAT in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| ACAT | |
| Polyclonal | |
| Western Blot 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 2-10 ug/ml lysate | |
| P35610 | |
| SOAT1 | |
| A fusion protein construct that contains the N-terminal region of human ACAT1. [UniProt# P35610] | |
| Unpurified | |
| RUO | |
| 6646 | |
| Human, Mouse, Hamster | |
| Antisera |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| ACACT, ACACT1, ACAT-1, ACATACAT1, acyl-Coenzyme A: cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase 1, Cholesterol acyltransferase 1, EC 2.3.1.26, RP11-215I23.2, STATSOAT, sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1, sterol O-acyltransferase 1 | |
| Rabbit | |
| 46 kDa | |
| 0.025 mL | |
| Primary | |
| This product is specific for protein corresponding to ACAT1 - Sterol O-acyltransferase 1 (SOAT1) Uniprot# P35610, Gene ID# 6646. [Important Note:- ACAT1 is a symbol for both acyl-Coenzyme A: cholesterol acyltransferase 1 (SOAT1/ACAT1) and for acetyl-CoA acetyltransferase 1 (ACAT1/ THIL, Gene ID 38, Uniprot P24752) which often cause confusion]. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction