missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ACAD11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | ACAD11 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066951
|
Novus Biologicals™
NBP3-30899-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066917
|
Novus Biologicals™
NBP3-30899-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACAD11 Polyclonal antibody specifically detects ACAD11 in Human samples. It is validated for Western BlotSpecifications
| ACAD11 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| Phosphate buffered saline | |
| 84129 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ACAD-11, acyl-CoA dehydrogenase family member 11, acyl-CoA dehydrogenase family, member 11, acyl-Coenzyme A dehydrogenase family, member 11, EC 1.3.99.-, FLJ12592, MGC150619 | |
| Synthetic peptide derived from the human ACAD11 protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title