missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ACAD11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30899-100ug
This item is not returnable.
View return policy
Description
ACAD11 Polyclonal antibody specifically detects ACAD11 in Human samples. It is validated for Western Blot
Specifications
| ACAD11 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| ACAD-11, acyl-CoA dehydrogenase family member 11, acyl-CoA dehydrogenase family, member 11, acyl-Coenzyme A dehydrogenase family, member 11, EC 1.3.99.-, FLJ12592, MGC150619 | |
| Synthetic peptide derived from the human ACAD11 protein | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 84129 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction