missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute Blue QPCR Mix, low ROX

ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments.

Brand:  Thermo Scientific™ AB4319A

Code : LE

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11832173

  • £451.00 / 400 reactions
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Thermo Scientific™ ABsolute Blue qPCR Mixes contain an inert blue dye to significantly enhance the contrast between reagent and plastic, allowing quick and easy verification that master mix has been dispensed.

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquoted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with different ROX passive reference dye concentrations
Specifications

Specifications

Contains:
• 1 x 5 mL ABsolute Blue QPCR Low ROX Mix (2X)
Sufficient for 400 x 25 μL reactions.

Store at -20°C
Tube
qPCR
Standard
ABsolute Blue
400 Reactions
2X
400 Reactions
5 mL
Primer-probe
High
Taq DNA Polymerase
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
qPCR Mix
Gene Expression
2 X
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.