missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, low ROX
ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments.
£451.00 - £1654.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11822173
|
Thermo Scientific™
AB4318B |
1,600 Reactions |
£1654.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11832173
|
Thermo Scientific™
AB4319A |
400 Reactions |
£451.00
400 reactions |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ ABsolute Blue qPCR Mixes contain an inert blue dye to significantly enhance the contrast between reagent and plastic, allowing quick and easy verification that master mix has been dispensed.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquoted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with different ROX passive reference dye concentrations
Specifications
Primer-probe | |
High | |
Taq DNA Polymerase | |
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P | |
qPCR Mix | |
2X | |
DNA, dsDNA |
Tube | |
qPCR | |
Standard | |
ABsolute Blue | |
Gene Expression | |
2 X |
For Research Use Only. Not for use in diagnostic procedures.