missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABO, Blood Group B Antigen Antibody (HEB-29), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£167.00 - £271.00
Specifications
| Antigen | ABO, Blood Group B Antigen |
|---|---|
| Clone | HEB-29 |
| Concentration | 1.0 mg/mL |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Aggregation |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Aggregation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275500
|
Novus Biologicals
NB120-2524 |
1 mL |
£271.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18504830
|
Novus Biologicals
NB120-2524SS |
0.25 ml |
£167.00
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABO, Blood Group B Antigen Monoclonal specifically detects ABO, Blood Group B Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Aggregation, Agglutination.Specifications
| ABO, Blood Group B Antigen | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Aggregation | |
| Unconjugated | |
| Mouse | |
| Human | |
| A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
| ABO | |
| IgM | |
| Concentrated Supernatant | |
| The antibody HEB-29 reacts with human blood group B. The specifity of the antibody HEB-29 was confirmed by comparison of specifity and reactivity to standard reagent using >5.000 samples of blood. |
| HEB-29 | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Aggregation | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| P16442 | |
| 28 | |
| Mixture of erythrocytes of blood group B and glycoprotein fraction isolated from the saliva of secretors with blood group B. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title