missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABO, Blood Group B Antigen Antibody (HEB-29), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB120-2524SS
This item is not returnable.
View return policy
Description
ABO, Blood Group B Antigen Monoclonal specifically detects ABO, Blood Group B Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Aggregation, Agglutination.
Specifications
| ABO, Blood Group B Antigen | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Aggregation | |
| A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
| Mouse | |
| Concentrated Supernatant | |
| RUO | |
| 28 | |
| Human | |
| Supernatant |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Aggregation | |
| HEB-29 | |
| Unconjugated | |
| P16442 | |
| ABO | |
| Mixture of erythrocytes of blood group B and glycoprotein fraction isolated from the saliva of secretors with blood group B. | |
| 0.25 ml | |
| Primary | |
| The antibody HEB-29 reacts with human blood group B. The specifity of the antibody HEB-29 was confirmed by comparison of specifity and reactivity to standard reagent using >5.000 samples of blood. | |
| Store at 4C. Do not freeze. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction