missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye California Red Phalloidin
Click to view available options
Quantity:
300 Reactions
Unit Size:
300 reactions
Description
Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Excitation (Max): 583nm
- Emission (Max): 605nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye California Red with Phalloidin conjugate. |
| For Use With (Application) | Conjugation |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye California Red Phalloidin |
| Sufficient For | 300 Reactions |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye California Red Phalloidin |
| Emission | 605 |
| Excitation | 583 |
For research use only (RUO).
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction