Learn More
Abnova™ Fluorescent Dye 700-I Streptavidin
Beschreibung
In comparison with Alexa Fluor 700, Fluorescence Dye 700-I are brighter with stronger absorption at 633nm. Fluorescence emission of Fluorescence Dye 700-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.
- Spectral Equivalent: Alexa Fluor 700
- Molar Extinction Co-efficient: ≥2,00,000 M-1cm-1
- Excitation (Max): 685nm
- Emission (Max): 710nm
- Solubility: Water
- Storage Buffer: Water
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Spezifikation
Spezifikation
| Description | Fluorescence Dye 700-I with Streptavidin conjugate. In comparison with Alexa Fluor 700, Fluorescence Dye 700-I are brighter with stronger absorption at 633nm. Fluorescence emission of Fluorescence Dye 700-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 200 μg |
| Product Type | Fluorescent Dye 700-I Streptavidin |
| Equivalence | Alexa Fluor 700 |
| Storage Buffer | Water |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C. |
| Label or Dye | Fluorescent Dye 700-I Streptavidin |
| Emission | 710 |
| Excitation | 685 |
For research use only (RUO).
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.