missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent Dye 680-I Streptavidin

Product Code. 16204960
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
This item is not returnable. View return policy

Product Code. 16204960

Brand: Abnova™ U0299.200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescence Dye 680-I with Streptavidin conjugate

Fluorescence emission of Fluorescence Dye 680-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.

  • Spectral Equivalent: Alexa Fluor 680, Cy5.5, IRDye 700
  • Molar Extinction Co-efficient: ≥2,00,000 M-1cm-1
  • Excitation (Max): 676nm
  • Emission (Max): 695nmnm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 680-I with Streptavidin conjugate. Fluorescence emission of Fluorescence Dye 680-I is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 200 μg
Product Type Fluorescent Dye 680-I Streptavidin
Equivalence Alexa Fluor 680, Cy5.5, IRDye 700
Storage Buffer Water
Content And Storage Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C.
Label or Dye Fluorescent Dye 680-I Streptavidin
Emission 695
Excitation 676

For research use only (RUO).

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.