Learn More
Abnova™ Fluorescent Dye 680-I Hydrazide
Description
Fluorescence emission of Fluorescent Dye 680-I Hydrazide is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 680, Cy5.5, IRDye 700
- Purity: >90%
- Molar Extinction Co-efficient: >2,00,000 M-1cm-1
- Excitation (Max): 676nm
- Emission (Max): 695nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable Schiff Base with carbonyl group. Fluorescence emission of Fluorescent Dye 680-I Hydrazide is well separated from that of other commonly used red fluorophores, such as TAMRA, Texas Red, Alexa Fluor 594, Fluorescent Dye 594-I, Fluorescent Dye 633-I, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Besides, their fluorescence is pH-independent from pH 3 to 11. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 680-I Hydrazide |
| Purity | >90% |
| Equivalence | Alexa Fluor 680, Cy5.5, IRDye 700 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 680-I Hydrazide |
| Emission | 695 |
| Show More |
For research use only (RUO).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.