missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent Dye 647-I Phalloidin

Product Code. 16294950
Change view
Click to view available options
Quantity:
300 Reactions
Unit Size:
300 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16294950 300 Reactions 300 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16294950 Supplier Abnova™ Supplier No. U0298.300Reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescence Dye 647-I with Phalloidin conjugate

The slight red shift in absorption spectrum makes Fluorescence Dye 647-I an optimal match to filters designed Cy5 dyes. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.

  • Spectral Equivalent: Alexa Fluor 647, Cy5, DyLight 650
  • Molar Extinction Co-efficient: ≥2,40,000 M-1cm-1
  • Excitation (Max): 649nm
  • Emission (Max): 664nm
  • Solubility: DMSO
  • Storage Buffer: DMSO

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 647-I with Phalloidin conjugate. The slight red shift in absorption spectrum makes Fluorescence Dye 647-I an optimal match to filters designed Cy5 dyes.Besides, their fluorescence is pH-independent from pH 3 to 11.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 300 Reactions
Product Type Fluorescent Dye 647-I Phalloidin
Sufficient For 300 Reactions
Equivalence Alexa Fluor 647, Cy5, DyLight 650
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 647-I Phalloidin
Emission 664
Excitation 649
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.