Learn More
Abnova™ Fluorescent Dye 555-I Phalloidin
Description
Compared with Cy3 conjugates, those of Fluorescence Dye 555-I are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Spectral Equivalent: Alexa Fluor 555, Cy3, DyLight 550, TRITC
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 555nm
- Emission (Max): 565nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 555-I with Phalloidin conjugate. Compared with Cy3 conjugates, those of Fluorescence Dye 555-I are slightly red-shifted, resulting in an optimal match to filters designed for Cy3 dyes in accompany with improved photostability. Besides, their fluorescence is pH-independent from pH 3 to 11. |
| For Use With (Application) | Conjugation |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye 555-I Phalloidin |
| Sufficient For | 300 Reactions |
| Equivalence | Alexa Fluor 555, Cy3, DyLight 550, TRITC |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 555-I Phalloidin |
| Emission | 565 |
| Show More |
For research use only (RUO).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.