missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye 532-I Streptavidin
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
Description
Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.
- Spectral Equivalent: Alexa Fluor 532
- Molar Extinction Co-efficient: ≥80,000 M-1cm-1
- Excitation (Max): 542nm
- Emission (Max): 558nm
- Solubility: Water
- Storage Buffer: Water
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 532-I with Streptavidin conjugate. |
| For Use With (Application) | Conjugation |
| Quantity | 200 μg |
| Product Type | Fluorescent Dye 532-I Streptavidin |
| Equivalence | Alexa Fluor 532 |
| Storage Buffer | Water |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C. |
| Label or Dye | Fluorescent Dye 532-I Streptavidin |
| Emission | 558 |
| Excitation | 542 |
For research use only (RUO).
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction