missing translation for 'onlineSavingsMsg'
Learn More

ABI3BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ABI3BP
Concentration 0.37 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16825795
View Documents
Proteintech
27727-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16815795
View Documents
Proteintech
27727-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ABI3BP contains 2 fibronectin type-III domains. The loss of ABI3BP expression could play a functional role in thyroid tumorigenesis. It also presumably represents a trigger gene for evoking cellular senescence, which has also been suggested to be involved in the prevention of tumorigenesis.
Specifications

Specifications

ABI3BP
Western Blot
Unconjugated
Rabbit
Mouse, Human
Q7Z7G0
25890, 320712
ABI3BP Fusion Protein Ag26841
Primary
-20°C
ABI3BP
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ABI3BP, Nesh binding protein, NESHBP, Target of Nesh SH3, TARSH
Abi3bp
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.