missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABI3BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27727-1-AP-20UL
This item is not returnable.
View return policy
Description
ABI3BP contains 2 fibronectin type-III domains. The loss of ABI3BP expression could play a functional role in thyroid tumorigenesis. It also presumably represents a trigger gene for evoking cellular senescence, which has also been suggested to be involved in the prevention of tumorigenesis.Specifications
| ABI3BP | |
| Polyclonal | |
| Unconjugated | |
| ABI3BP | |
| ABI3BP, Nesh binding protein, NESHBP, Target of Nesh SH3, TARSH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25890, 320712 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z7G0 | |
| Abi3bp | |
| ABI3BP Fusion Protein Ag26841 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction